Learn More
Eppendorf™ Rotor Lids
Designed for use with Eppendorf Rotors
Brand:  Eppendorf™ 5409707000
Code : NEW
Additional Details : Weight : 0.19800kg
 View more versions of this product
Product Code. 15244536
  • 265.00 € / Each
In Stock
to see stock.
Description
Description
  • Replacement rotor lids
  • Aerosol-tight and aluminum lids available
  • Available in a variety of sizes
Specifications
Specifications
Rotor Lid
Aluminum
Centrifuge 5427R, FA–45-30-11
Aerosol-tight