Learn More
Fritsch™ Impact Rotors for PULVERISETTE 14
Description
- Extremely durable, low-wear stainless steel
- Impact rotors with 8-, 12-, 24-ribs
- For the pulverisation of medium-hard, soft, brittle and fibrous substances - from lime to plants
Specifications
Specifications
| Type | Rotor |
| For Use With (Equipment) | PULVERISETTE 14 Variable Speed Rotor Mill, for fast comminution of fibrous substances |
| No. of Places | 24 Ribs |