missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Fixed-angle lighter rotor for harvesting of bacteria, cells or yeast and removing cellular debris for e.g. recombinant protein or cell compartment purification
- Includes 6 x 250 polypropylene bottle
- Max. speed: 26,400 x g (13,000 rpm)
- Max. capacity: 6 x 250 mL (Nominal capacity)
- Made from high-quality aluminum
Specifications
Specifications
| Material | Aluminum |
| Type | Fixed Angle Rotor |
| Capacity | 6 x 250 mL |
| Max. RCF | 26,400 x g |
| Max. Speed | 13,000 rpm |
| For Use With (Equipment) | Centrifuge CR22N/CR21N |
| Biocontainment | No |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?