missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Rotor for PCV Series Compact Benchtop Centrifuges

Product Code. 11481028
Change view
Click to view available options
No. of Places:
16
16 (2 microtube strips)
18
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places unitSize
11481028 16 (2 microtube strips) Each
11451028 16 Each
11471028 18 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481028 Supplier Grant Instruments™ Supplier No. PSR16

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fits PCV-2400 Combi-Spin and PVC6400 Multi-spin benchtop centrifuges. Grant Intstruments™ Rotor for PCV Series Compact Benchtop Centrifuges provide versatility with interchangeable rotors for various microtube sizes.

  • PR2-05: Interchangeable centrifuge rotor for 8 x 1.5/2.0mL plus 8 x 0.5mL microtubes (optional)
  • PR2-05-02 : Interchangeable centrifuge rotor for 6 x 1.5/2.0mL plus 6 x 0.2mL microtubes (optional)
  • PSR-16: Interchangeable centrifuge rotor for 2 x 8-well 0.2mL microtube strips (optional)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 2 x 8-well 0.2 mL Microtube Strips
No. of Places 16 (2 microtube strips)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.