missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rottlerin

Product Code. 30001271
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001271 10 mg 10mg
18024172 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001271 Supplier R&D Systems™ Supplier No. 1610/10
  • ONLINE EXCLUSIVE PRICE
    157.55 €
    141.75 € / 10mg
    Save 15.80 €
    10% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Reported PKCδ inhibitor

Originally reported to inhibit PKC isoforms. Also reported to inhibit CAM kinase III. However, recently shown to inhibit a wide range of protein kinases, and most potently to inhibit PRAK and MAPKAP-K2 (IC50 values are 1.9 and 5 μM respectively). Also shown to act as a direct mitochondrial uncoupler. Thought to stimulate autophagy by targeting upstream mTORC1 control pathways.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-08-6
Molecular Formula C30H28O8

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 516.55
Purity >98%
Molecular Weight (g/mol) 516.546
Chemical Name or Material Rottlerin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.