missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Round Bottom Flasks

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07035028

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10030371

  • 133.00 € / Pack of 10
Estimated Shipment: 26-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications

Specifications

34 mm
50 mL
Duran Glass
105 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers