missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT Round Canisters
Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
2 L
4 L
5 L
10 L
25 L
50 L
60 L
Closure Size:
53B
83B
S65
Unit Size:
Each
Description
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 L to 4 L, 10 L, 25 L, 50 L and 60 L for the collection of voluminous solvents
Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances.
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 10 L |
| Closure Size | 83B |
| Color | White (Translucent) |
| Description | Round canister 10 L, B83, PP, with carrying handles, dimensions (W x H x D): 250 x 390 x 250 mm, autoclavable |
| Dimensions (L x W x H) | 250 x 250 x 390 mm |
| Disposable | Reusable |
| Includes | Carrying handles, autoclavable |
| Material | Polypropylene |
| Mouth Style | B83 Thread |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction