missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT Round Canisters
Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
2 L
4 L
5 L
10 L
25 L
50 L
60 L
Closure Size:
53B
83B
S65
Unit Size:
Each
Description
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 L to 4 L, 10 L, 25 L, 50 L and 60 L for the collection of voluminous solvents
Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances.
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
Tekniske data
Tekniske data
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 10 L |
| Closure Size | S65 |
| Color | White (Translucent) |
| Description | Round canister 10 L, S65, PE-HD, dimensions (W x H x D): 205 x 430 x 205 mm |
| Dimensions (L x W x H) | 205 x 205 x 430 mm |
| Disposable | Reusable |
| Includes | Handle and measuring scale |
| Material | HDPE |
| Mouth Style | S65 Thread |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion