Learn More
Round Glass Narrow Mouth Alpha Bottles, without Cap
Product Code. 12810904
Change view
Click to view available options
Capacity (Metric):
125 mL
Color:
Clear
Unit Size:
Pack of 104
Product Code. Capacity (Metric) Color Unit Size
12810904 125 mL Clear Pack of 104
1 options
Product Code. 12810904 Supplier SGD PHARMA Supplier No. 300735
  • ONLINE EXCLUSIVE PRICE
    28.85 €
    28.37 € / Pack of 104
    Save 0.48 €
    1% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Store, pour or share your laboratory liquids using Round Glass Narrow Mouth Alpha Bottles, without Cap. Made from amber or clear borosilicate glass, the round bottles are an integral tool in the laboratory.
  • Narrow neck
  • Borosilicate glass
  • Round
Specifications
Capacity (Metric) 125 mL
Color Clear
Material Borosilicate Glass
Closure Liner Polypropylene/SOP
Closure Material Polypropylene
Product Type Alpha Bottle