missing translation for 'onlineSavingsMsg'
Learn More

Foxx Life Sciences Round PP Vacuum Trap System

Product Code. 31933498
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
2 L
2 L (Primary), 1 L (Overflow)
4 L
4 L (Primary), 1 L (Overflow)
4 L (Primary), 2 L (Overflow)
4 L (Primary), 4 L (Overflow)
Unit Size:
1 set
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31933498 2 L Each
31933366 2 L (Primary), 1 L (Overflow) 1 set
31933359 4 L Each
31933495 4 L (Primary), 1 L (Overflow) 1 set
31933420 4 L (Primary), 2 L (Overflow) 1 set
31933382 4 L (Primary), 4 L (Overflow) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31933498 Supplier Foxx Life Sciences Supplier No. 3052003FLS
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to meet strict CDC laboratory guidelines for handling biohazardous waste

  • Features:
    • Vacuum trap assembly with autoclavable PP bottles
    • 40 CFR 264.175 compliant aspirator system has a compact, tip-resistant secondary container for stability
    • Wide mouth PP bottles allow easy and thorough cleaning
    • Standard vacuum trap with 53B VersaCap™ and 83B VersaCap™ closure system with rotating adapter prevents tube tangling
  • Applications
    • Aspiration of cell culture media
    • Vacuum pump trap kit
    • Waste collection system for high-purity processes
    • Ideal for house vacuum or vacuum pump trap systems
    • Aspirator bottle assembly

Specifications

Capacity (Metric) 2 L
Case Material Polypropylene
Material Polypropylene
Maximum Pressure 1 bar
Includes Secondary Container, Bottle, Cap Assembly with Vent Filter, Tubing, Pinch Clamp
Length (Metric) 228.6 mm
Type Round PP Vacuum Trap System
Width (Metric) 279.4 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.