missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Routine DNA Ladder

Product Code. 10284633
Change view
Click to view available options
No. of Bands:
11
16
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Bands unitSize
10284633 11 Each
10450464 16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10284633 Supplier Fisher BioReagents™ Supplier No. BP2581200
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ready-to-use DNA size standard is room-temperature stable for up to two years

  • Includes a landmark doublet at 1400 and 1550 bp

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Form Liquid
For Use With (Application) PCR size confirmation
No. of Bands 11
Quantity 1000 μL
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.