missing translation for 'onlineSavingsMsg'
Learn More

Roxatidine Acetate Hydrochloride, MedChemExpress

Product Code. 16460868
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460868 5 g 5g
16452129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460868 Supplier Medchem Express Supplier No. HYB0305A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Roxatidine Acetate Hydrochloride is a potent, selective, competitive and orally active histamine H2-receptor antagonist. Roxatidine Acetate Hydrochloride has antisecretory potency against gastric acid secretion. Roxatidine Acetate Hydrochloride can also suppress inflammatory responses and can be used for gastric and duodenal ulcers research. Roxatidine Acetate Hydrochloride has antitumor activity.

Roxatidine Acetate Hydrochloride is a potent, selective, competitive and orally active histamine H2-receptor antagonist. Roxatidine Acetate Hydrochloride has antisecretory potency against gastric acid secretion. Roxatidine Acetate Hydrochloride can also suppress inflammatory responses and can be used for gastric and duodenal ulcers research. Roxatidine Acetate Hydrochloride has antitumor activity.

Specifications

Chemical Name or Material Roxatidine Acetate Hydrochloride
CAS 93793-83-0
Color White
Purity Grade Notes Research
Molecular Formula C19H29ClN2O4
Quantity 5 g
Synonym HOE 760
Solubility Information DMSO : 50 mg/mL (129.90 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (129.90 mM)
SMILES O=C(NCCCOC1=CC=CC(CN2CCCCC2)=C1)COC(C)=O.Cl
Molecular Weight (g/mol) 384.9
Formula Weight 384.9
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.