Learn More
Emma Safety Footwear Roy XD Safety Shoes
Product Code. 15968851
Change view
Click to view available options
Size: 49
Product Code.
15968851 Size: 49
1 options
Product Code. 15968851 Supplier Emma Safety Footwear Supplier No. MM70156049
  • ONLINE EXCLUSIVE PRICE
    110.25 €
    69.62 € / 1 pairs
    Save 40.63 €
    36% Off
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Versatile shoe with wipe clean uppers and an 'easy clean' sole. The shoe sculpts itself to the wearer's foot for comfort while an internal toe cap provides resilient protection against falling objects and unforeseen hazards.
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver
Specifications
Design Roy XD
Material Leather
Size 49