Learn More
uvex RPL Hypervision CB Clear AF
Product Code. 31751173 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751173 Polycarbonate Each
1 options
Product Code. 31751173 Supplier uvex Supplier No. 9321955
  • ONLINE EXCLUSIVE PRICE
    11.44 €
    10.96 € / Pack of 1
    Save 0.48 €
    4% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
For reliable visual clarity and protection.
  • RPL Hypervision CB eyewear featuring a clear AF lens, designed to deliver dependable visual clarity and consistent performance, identified as Hypervision CB clear AF.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Replacement Lens
Disposable Reusable
Lens Material Polycarbonate
Autoclavable Not Autoclavable