Learn More
uvex RPL Hypervision PC Replacement Lens - Clear SV EXC
Product Code. 31751172 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751172 Polycarbonate Each
1 options
Product Code. 31751172 Supplier uvex Supplier No. 9321255
  • ONLINE EXCLUSIVE PRICE
    19.12 €
    18.31 € / Pack of 1
    Save 0.81 €
    4% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement lens for RPL Hypervision safety spectacles, enabling sustainable use by extending the product life.
Act sustainably and use our replacement lenses to extend the life of your safety spectacles. Scratched lens? No problem, simply replace the lens and keep wearing the same safety spectacles.
  • Replacement lens designed for RPL Hypervision safety spectacles, allowing worn or scratched lenses to be easily replaced to extend the service life of the eyewear as a sustainable solution.
  • Clear SV EXC polycarbonate lens offering an enlarged field of vision for optimal visibility of hazards while maintaining compatibility with the Hypervision system.
  • Certified in accordance with EN ISO 16321-1 for personal eye and face protection for industrial use, including UV protection filtering.
  • Robust polycarbonate lens featuring proven uvex supravision coating technology with permanent anti-fog and scratch-resistant properties.
  • UV400 protection providing 100% protection against hazardous ultraviolet radiation up to 400 nm.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Replacement Lens
Disposable Reusable
Lens Material Polycarbonate
Autoclavable Not Autoclavable