missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Composed of recombinant protein A coupled with cross-linked 4% agarose base matrix
- High dynamic binding capacity, single-point attachment of protein A, with high recovery, high purity and low ligand leakage
- No mammalian components involved during manufacturing process
- Matrix: 4% cross-linked agarose
- Average particle size: 90 μm (d50V)
- Ligand: 6 mg rProtein A/mL medium (E. coli)
- Binding capacity: approx. 50 mg human IgG/mL media
- pH stability working range: 3 to 10
- Pressure/Flow specification: base matrix 150 to 250 cm/hr., 100 kPa, XK 50/60 column, bed
Specifications
Specifications
| Quantity | 5 mL |
| Stability | Stable in aqueous Buffers commonly used in Protein A Chromatography |
| For Use With (Application) | For PurifiCatIon of Monoclonal and Polyclonal Antibodies at Labs and Process Scales |
| Storage Requirements | 2°C to 8°C |
| Binding Capacity | 50 mg Human lgG/mL medium2 |
| Product Type | rProtein A Sepharose Fast Flow Media |
| Flow Rate | 150 to 250 cm/Hoμr |
| Particle Size | 60 to 165 μm |
| Height (English) Bed | 9.84 in. |
| Height (Metric) Bed | 25 cm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?