missing translation for 'onlineSavingsMsg'
Learn More

RR-11a analog, MedChemExpress

Product Code. 16484598
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484598 10 mg 10mg
16480897 50 mg 50mg
16475446 100 mg 100mg
16437317 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484598 Supplier Medchem Express Supplier No. HY112205A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RR-11a analog is a potent and irreversible inhibitor of Schistosoma mansoni legumain, with an IC50 of 31 nM. RR-11a analog is an aza-Asn derivative and aza-peptide Michael acceptor.

RR-11a analog is a potent and irreversible inhibitor of Schistosoma mansoni legumain, with an IC50 of 31 nM. RR-11a analog is an aza-Asn derivative and aza-peptide Michael acceptor.

Specifications

Chemical Name or Material RR-11a analog
CAS 685543-66-2
Color White
Purity Grade Notes Research
Molecular Formula C22H29N5O8
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (254.33 mM; Need ultrasonic)
SMILES O=C(N[C@@H](C)C(N[C@@H](C)C(NN(CC(N)=O)C(/C=C/C(OCC)=O)=O)=O)=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 491.49
Formula Weight 491.49
Percent Purity 98.79%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.