missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RS-1
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Description
- RS-1 is an enhancer of RAD51, an enzyme that assists in the homology-directed repair (HDR) pathway of DNA double-strand breaks, and is reported to function by stabilizing the active form of RAD51 and by enhancing recombinogenic activity of the enzyme (Jayathilaka et al.) Via this mechanism, RS-1 can stimulate HDR, thereby enhancing precise HDR-dependent CRISPR-Cas9 genome editing in some cell lines (Pinder et al.; Song et al.)
- Promotes HDR, which is reported to enhance knock-in efficiency of CRISPR-Cas9 genome editing (Pinder et al.; Song et al.; however, these effects are cell type-specific and context-dependent (Xie et al.; Zhang et al.)
- Stimulates the DNA-binding activity of RAD51 to promote cancer cell death and induce anti-tumor responses in a mouse xenograft tumor model (Mason et al.)
Specifications
Specifications
| CAS | 312756-74-4 |
| Quantity | 10 mg |
| Molecular Formula | C20H16Br2N2O3S |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction