missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RS-1

Product Code. p-7217435
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 17170322

Brand: STEMCELL Technologies 74092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HDR pathway enhancer; Enhances RAD51

  • RS-1 is an enhancer of RAD51, an enzyme that assists in the homology-directed repair (HDR) pathway of DNA double-strand breaks, and is reported to function by stabilizing the active form of RAD51 and by enhancing recombinogenic activity of the enzyme (Jayathilaka et al.) Via this mechanism, RS-1 can stimulate HDR, thereby enhancing precise HDR-dependent CRISPR-Cas9 genome editing in some cell lines (Pinder et al.; Song et al.)
  • Promotes HDR, which is reported to enhance knock-in efficiency of CRISPR-Cas9 genome editing (Pinder et al.; Song et al.; however, these effects are cell type-specific and context-dependent (Xie et al.; Zhang et al.)
  • Stimulates the DNA-binding activity of RAD51 to promote cancer cell death and induce anti-tumor responses in a mouse xenograft tumor model (Mason et al.)

Specifications

CAS 312756-74-4
Quantity 10 mg
Molecular Formula C20H16Br2N2O3S
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.