missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RS 127445 hydrochloride
GREENER_CHOICE

Product Code. 30001336
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001336

Brand: Tocris Bioscience™ 2993/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and selective 5-HT2B antagonist

High affinity 5-HT2B receptor antagonist (pKi = 9.5). Displays 1000-fold selectivity for 5-HT2B with good bioavailability.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199864-86-3
Molecular Formula C17H17ClFN3

Specifications

Quantity 10 mg
Formula Weight 317.79
Purity >98%
Molecular Weight (g/mol) 317.79
Chemical Name or Material RS 127445 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.