missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ RS 17053 hydrochloride
Description
α1A-adrenoceptor antagonist, with very high affinity for α1A receptors (pKi and pA2 estimates of 9.1 - 9.9) and a 30 - 100 fold selectivity over the α1B and α1D subtypes (pKi and pA2 estimates 7.7 - 7.8).
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 449.42 |
| Purity | >98% |
| Molecular Weight (g/mol) | 449.416 |
| Chemical Name or Material | RS 17053 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?