missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ RS 232 Cable (9-pole)

Product Code. 15738049 Shop All Heidolph Instruments Products
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15738049 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15738049 Supplier Heidolph™ Supplier No. 1400704072
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cable for use with Hei-Connect or Hei-TORQUE Ultimate models.

  • Supplies power to Ultimate magnetic and overhead stirrers
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Equipment) Heidolph™ Hei-Connect or Hei-TORQUE Ultimate Stirrers
Type 9-pole Cable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.