Learn More
RS Components RS PRO INVERTIBLE AIR DUSTER
Brand:  RS Components 168-1646
Encompass
RS PRO INVERTIBLE AIR DUSTER
Product Code. 15920727
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • RS invertable dust remover
Signal Word
  • Warning
Hazard Category
  • Flammable and non-flammable aerosols category 3
Hazard Statement
  • H229-Pressurised container: May burst if heated.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P251-Do not pierce or burn, even after use.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.