missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (RS)-(±)-Sulpiride

Product Code. 30001141
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001141 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001141 Supplier R&D Systems™ Supplier No. 0894/100
  • ONLINE EXCLUSIVE PRICE
    126.86 €
    113.40 € / 100mg
    Save 13.46 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Standard selective D2-like antagonist

Standard D2-like dopamine receptor antagonist.

S-enantiomer also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15676-16-1
Quantity 100 mg
Product Line Tocris
Molecular Formula C15H23N3O4S
Formula Weight 341.42
Purity >99%
Molecular Weight (g/mol) 341.43
Chemical Name or Material (RS)-(±)-Sulpiride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.