Learn More
RS232 Interface Kit, for OHAUS™ Scout STX and SKX Precision Portable Balances
Product Code. 15589631 Shop All Ohaus Products
Click to view available options
:
RS232 Interface Kit
Product Code. 15589631
Brand: Ohaus 30268982
For use with Scout STX and SKX series Precision Portable Balances.
RS232 Interface Kit cable terminates with a 9 pin Sub-D female connector.
TRUSTED_SUSTAINABILITY
Specifications
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Product Line Interface Cables and Kits