missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ rU Phosphoramidite, standard grade, serum vial bottle

Product Code. 17977182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
Product Code. Quantity unitSize
17977182 2 g 2g
1 options
This item is not returnable. View return policy

Product Code. 17977182

Brand: Thermo Scientific™ 27140407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

rU CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) for use in oligonucleotide synthesis.

rU CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups (except T and U) designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-O-TBDMSi-uridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C45H61N4O9PSi
Base: rU
Formula weight: 861.1 g/mol
CAS number: 118362-03-1
HPLC Purity: ≥98.0%
31P NMR Purity: ≥98.0%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type rU CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Uridine (U)
Shipping Condition Room Temperature
Purity 98%
Quantity 2 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.