Learn More
Abeba™ Rubber 5300 Shoes
Product Code. 15611898
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15611898
Brand: Abeba™ 5300/44
Only null left
Add to basket
Add to basket
ESD dissipative shoe is acid and base resistant.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • White
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Rubber
Style 5300