Learn More
Abeba™ Rubber 9252 Shoes
Product Code. 15603218
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15603218 36 1 pair
15613218 37 1 pair
15623218 38 1 pair
15633218 39 1 pair
15643218 40 1 pair
15653218 41 1 pair
15663218 42 1 pair
15673218 43 1 pair
15683218 44 1 pair
15693218 45 1 pair
15603228 46 1 pair
15613228 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15603218 Supplier Abeba™ Supplier No. 9252/36
Only null left
Add to basket
Add to basket
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC/li
Color Black
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Rubber
Style 9252