Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Product Code. 11870950
Change view
Click to view available options
Color:
Red
Unit Size:
Each
Product Code. Color Unit Size
11870950 Red Each
1 options
Product Code. 11870950 Supplier Deutsch Neumann™ Supplier No. 4719901
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ The Flip™ Safety Pipette Fillers are manufactured from natural rubber.
  • Only two operating points
  • Evacuate via the new automatic valve
  • One-handed design is the simplest safety pipette filler available
  • Ability to clean inside of bulb by removing patented valve and rinsing out
Specifications
Color Red
Material Rubber
Type Flip Security Pear Pipette
Quantity 1 Each