missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rubber Mats

For use with Eppendorf Adapters and buckets

Brand:  Eppendorf™ 5702708003

Code : 99

Additional Details : Weight : 1.05000kg

 View more versions of this product

Product Code. 10208121

  • 26.40 € / Pack of 10
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Rubber mats
  • Available for a variety of different rotors and adapters
Specifications

Specifications

Mat
Rotor F-35-30-17
1 x 15 mL Round Bottom Tube
Set of 10 mats
Rubber
Black
For 1 round-bottom tubes 15mL, for Rotor F-35-30-17, 10 pieces
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers