Learn More
Manutan Rubber ribbed mobile step, yellow, three step with handle
Description
Thick ribbed rubber treads for secure foot holds. Formed sheet steel steps. Specially designed steel domed feet with rubber protection ring that grips the floor when in use. 50mm diameter spring loaded retractable castors with non-marking tyres. Feet bolted to legs for easy replacement. Hard wearing powder coated paint finish.
- Max. load capacity: 150kg
- Number of steps (platform included): 3
- With handrail
- Platform height: 765mm
- O/A Dimensions (H x W x D): 1425mm x 564mm x 784mm
- Yellow
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.