missing translation for 'onlineSavingsMsg'
Learn More

Marienfeld Superior™ Rubber Safety Pipette Filler

Product Code. 10781294
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10781294 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10781294 Supplier Marienfeld Superior™ Supplier No. 5900001
Stem Cell Research Savings
Use promo code 28032
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this Marienfeld Superior™ Rubber Safety Pipette Fillers for graduated and volumetric pipettes

  • Made of red rubber (natural caoutchouc)
  • For graduated and volumetric pipettes
  • Control of the functions by squeezing the appropriate valves between thumb and forefinger
  • Individually packed in foil with short instruction for use

Specifications

Color Red
Material Natural Rubber
Model Universal
Type Pipette Filler
For Use With (Equipment) Volumetric and graduated pipettes
Functional Range Up to 100 ml capacity and 5-9 mm diameter
Quantity 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.