missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ RuBi-GABA

Product Code. 18023972
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18023972 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18023972 Supplier R&D Systems™ Supplier No. 3400/10
  • ONLINE EXCLUSIVE PRICE
    346.84 €
    309.75 € / 10mg
    Save 37.09 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Caged GABA; excitable at visible wavelengths

Ruthenium-bipyridine-triphenylphosphine caged GABA (Cat. No. 0344) that is excited by visible wavelengths. Provides greater tissue penetration, less phototoxicity, faster photorelease kinetics and better spatial resolution than UV light-sensitive caged compounds. Produces GABA receptor-mediated currents in pyramidal neurons in vitro and displays no effect on endogenous GABAergic or glutamatergic transmission at concentrations effective for uncaging.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1028141-88-9
Quantity 10 mg
Product Line Tocris
Molecular Formula C42H39F6N5O2P2Ru
Formula Weight 922.8
Purity >98%
Molecular Weight (g/mol) 922.815
Chemical Name or Material RuBi-GABA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.