missing translation for 'onlineSavingsMsg'
Learn More

Rubidium Chloride, Extra Pure, SLR, Fisher Chemical

Product Code. 10549390 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10549390 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10549390 Supplier Fisher Chemical Supplier No. R/0500/44
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 1390°C
Melting Point 718°C
CAS 11-9-7791
Color White
Physical Form Solid
Quantity 5 g
Molecular Formula ClRb
MDL Number MFCD00011187
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES [Cl-].[Rb+]
IUPAC Name rubidium(1+);chloride
Molecular Weight (g/mol) 120.92
PubChem CID 62683
ChEBI CHEBI:78672
Formula Weight 120.92g/mol
Packaging Glass Jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.