Learn More
Rubidium formate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals
Descrizione
Rubidium formate hydrate is used in anticoagulants, dental materials, indicators and reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Color | White |
| Linear Formula | RbOOCH·xH2O |
| Quantity | 50 g |
| Sensitivity | Hygroscopic |
| Formula Weight | 130.49 (Anhydrous) |
| Percent Purity | 99.8% |
| Physical Form | Crystalline |
| Assay | (metals basis) |
| Chemical Name or Material | Rubidium formate hydrate |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.