missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex NB60SZ Chemical Protection Gloves

Product Code. 17290242 Shop All Uvex Products
Change view
Click to view available options
Size:
9
10
11
Unit Size:
1 pair
10 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17290242 10 10 pairs
17200411 9 10 pairs
17210411 11 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17290242 Supplier uvex Supplier No. 9893061
  • ONLINE EXCLUSIVE PRICE
    457.00 €
    425.73 € / 10 pairs
    Save 31.27 €
    6% Off
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Full NBR-coating reinforced with cotton interlock supporting material for applications that require protection against chemicals and mechanical risks. Elastic on the glove cuff provides additional protection by preventing hazards from entering the glove.

  • Extended length chemical protection glove with reinforced cotton interlock lining
  • Cotton liner helps absorb water vapor
  • Additional elastic collar at gauntlet end
  • Resistant to many chemicals, acids, alkalis, mineral oils, and solvents
  • Suitable for applications that require protection against chemicals and mechanical risks (ex. heat)
TRUSTED_SUSTAINABILITY

Specifications

Cuff Material Elastic
Applications Laboratory
Coating Material NBR
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 10
EN388 Standard 2121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKOPT
For Use With (Application) Chemical Handling, Drainage Construction, and Municipal Cleaning
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Rubiflex NB60SZ
Length (Metric) 60 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.