missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Rubiflex Protective Gloves

Product Code. 15398076 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
37 cm
60 cm
Glove Cuff Style:
Long Guantlet
Short Gauntlet
Unit Size:
1 pair
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Glove Cuff Style unitSize
15398076 37 cm Short Gauntlet 1 pair
11971641 60 cm Long Guantlet 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Attention! Make sure to pay attention to the List of Chemical Resistance!
Product Code. 15398076 Supplier Buerkle™ Supplier No. 25070002
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Attention! Make sure to pay attention to the List of Chemical Resistance!

Rubiflex protective gloves – with short or long gauntlet. Safe working with fats, mineral oils, solvents and many acids and alkalis.

  • Protective gloves with short or long gauntle
  • For fats, mineral oils, solvents and many acids and alkalis
  • Made from nitrile rubber
  • Very flexible, soft to the touch
  • High abrasion and cut resistance
  • Comfortable cotton interlock interior
  • To standard EN 374
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 374
Color Green
Material Nitrile Rubber
Product Type Protective Gloves
For Use With (Application) Handling Fats, Mineral Oils, Solvents, Many Acids and Alkalis
Glove Cuff Style Short Gauntlet
Lining Cotton
Resistant To Abrasion and cuts
Length (Metric) 37 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.