missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex S NB27S Chemical Protection Gloves

Product Code. 17270242 Shop All Uvex Products
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17270242 9 10 pairs
17230242 8 10 pairs
17240252 10 10 pairs
17240242 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17270242 Supplier uvex Supplier No. 9893059
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    149.25 € / 10 pairs
    Save 1.75 €
    1% Off
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are fully coated with NBR for chemical resistance, and feature a high-quality cotton interlock liner that provides heat insulation for increased mechanical protection.

  • NBR-coated safety gloves with reinforced cotton interlock supporting material
  • Thick, reinforced liner provides insulation for thermal protection
  • Resistant to a variety of chemicals, acids, alkalis, mineral oils, and solvents
  • Suitable for applications that require protection against chemicals and various mechanical risks
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material NBR
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 9
EN388 Standard 2121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKNOPT
For Use With (Application) Automotive, Chemical Handling, Food Processing, Machine Operation, Metal Work, and Sandblasting
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Rubiflex S NB27S
Length (Metric) 27 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.