missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex S NB35S Chemical Protection Gloves

Product Code. 15383453 Shop All Uvex Products
Change view
Click to view available options
Size:
9
11
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15383453 11 Each
15303463 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15383453 Supplier uvex Supplier No. 9889123
  • ONLINE EXCLUSIVE PRICE
    16.80 €
    15.09 € / Pack of 1
    Save 1.71 €
    10% Off
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cotton interlock supporting material, along with NBR exterior, makes these gloves suitable for applications that require protection against chemicals and mechanical hazards.

  • Long NBR chemical protection gloves reinforced cotton interlock carrier material
  • NBR coating provides mechanical abrasion resistance
  • High-quality interlock cotton supporting material increases wearer comfort
  • Gloves are resistant to a wide variety of chemicals, acids, alkalis, mineral oils, and solvents
  • Ergonomic fit with improved dexterity and instinct
  • Ideal for applications in the chemical industry, sewer construction work, and city cleaning
TRUSTED_SUSTAINABILITY

Specifications

Coating Material NBR
Color Green
Hand Specific
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 11
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKNOPT
Glove Cuff Style Gauntlet
Liner Material Interlock Cotton
Series Rubiflex S NB35S
Length (Metric) 35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.