Learn More
uvex Rubiflex S NB35SF Safety Glove
Product Code. 17797609 Shop All Uvex Products
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17797609 9 10 pairs
17787609 8 10 pairs
17707619 10 10 pairs
17717619 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17797609 Supplier uvex Supplier No. 6020909
  • ONLINE EXCLUSIVE PRICE
    289.00 €
    282.98 € / 10 pairs
    Save 6.02 €
    2% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The comfortable NBR safety glove, seamlessly coated in a reinforced design, with chemical and abrasion resistance.
  • Good heat/cold insulation on the palm
  • Abrasion and cut resistance
  • Ergonomic fit
  • High flexibility
  • Excellent mechanical characteristics
  • Good resistance to many chemicals, acids, alkalis, mineral oils and solvents
  • High water vapor absorption due to the cotton lining
  • Fully coated
  • Sandwich lining on palm, cuff
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN: 388 (2 1 2 1 X)
Color Green
Latex Free Yes
Material Cotton, Fleece, NBR
Product Type Chemical-resistant Gloves
Size 9