missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex S NB60S Chemical Protection Gloves

Product Code. 17280242 Shop All Uvex Products
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
1 pair
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17280242 10 10 pairs
17250242 8 1 pair
17260242 9 10 pairs
17200252 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17280242 Supplier uvex Supplier No. 9893060
  • ONLINE EXCLUSIVE PRICE
    387.00 €
    360.61 € / 10 pairs
    Save 26.39 €
    6% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Long, NBR-coated chemical protection gloves feature reinforced cotton interlock supporting material for applications that require protection against chemicals and mechanical hazards.

  • NBR-coated chemical protection gloves with an extended gauntlet cuff
  • Reinforced cotton interlock supporting material provides mechanical protection
  • Resistant to many chemicals, acids, alkalis, mineral oils, and solvents
  • Ergonomic design is comfortable and highly flexible
  • Silicone-free according to imprint test
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material NBR
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 10
EN388 Standard 2121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKOPT
For Use With (Application) Chemical Handling, Municipal Cleaning, Sandblasting, and Sewer Construction
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Rubiflex S NB60S
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.