Learn More
uvex Rubiflex S NB80S Chemical Protection Gloves
Product Code. 15356952 Shop All Uvex Products
Change view
Click to view available options
Size:
9
10
11
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15356952 10 Each
15376952 9 Each
15366952 11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15356952 Supplier uvex Supplier No. 6019002
Stock Up Your Lab
Use promo code 28007
View all promotions
In Stock
Add to basket
Edge
Add to basket
Cotton interlock supporting material, along with NBR exterior, makes these gloves suitable for applications that require protection against chemicals and mechanical hazards.
  • Long NBR chemical protection gloves reinforced cotton interlock carrier material
  • NBR coating provides mechanical abrasion resistance
  • High-quality interlock cotton supporting material increases wearer comfort
  • Gloves are resistant to a wide variety of chemicals, acids, alkalis, mineral oils, and solvents
  • Ergonomic fit with improved dexterity and instinct
  • Ideal for applications in the chemical industry, sewer construction work, and city cleaning
TRUSTED_SUSTAINABILITY
Specifications
Coating Material NBR
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 10
EN388 Standard 2121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKOPT
Glove Cuff Style Gauntlet
Liner Material Interlock Cotton
Series Rubiflex S NB80S
Length (Metric) 80 cm