missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex S XG35B Chemical Protection Gloves

Product Code. 15337112 Shop All Uvex Products
Change view
Click to view available options
Size:
7
10
Unit Size:
10 pairs
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15337112 10 Each
17290441 7 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15337112 Supplier uvex Supplier No. 6055710
  • ONLINE EXCLUSIVE PRICE
    16.90 €
    15.21 € / Pack of 1
    Save 1.69 €
    10% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight, stockinette NBR chemical protection gloves feature Xtra Grip technology for enhanced grip in wet or oily areas.

  • Lightweight, stockinette NBR chemical protection gloves with XG Grip coating
  • Xtra Grip technology provides a secure grip in wet and oily areas
  • Cotton interlock supporting material ensures a high level of user comfort
  • Multi-layered construction provides mechanical abrasion resistance
  • Resistant to grease, mineral oils, and a wide variety of chemicals
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Coating Material NBR
Color Blue, Black
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 10
EN388 Standard 3121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKNOPT
For Use With (Application) Automotive, Chemical Handling, Laboratory Work, Maintenance, and Metal Handling
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Rubiflex S XG35B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.