missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubipor XS2001 Safety Gloves

Product Code. 17240421 Shop All Uvex Products
Change view
Click to view available options
:
5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17240421 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17240421 Supplier uvex Supplier No. 6027605
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight, breathable gloves feature a thin layer of NBR-impregnation that has been measured and confirmed using the Hohenstein Institute's skin model.

  • Lightweight, elastic safety gloves with NBR-impregnation
  • Highly flexible stretch cotton backing material with elastane
  • Gloves are extremely breathable, measured and confirmed using the Hohenstein Institute's skin model
  • Suitable for operating touch displays
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material NBR
Color White
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Cotton, Elastane
Product Type Mechanical Protection Gloves
Size 5
EN388 Standard 1110X
For Use With (Application) Assembly, Inspection, Precision Work, and Sorting
Gender Unisex
Glove Cuff Style Knit
Series Rubipor XS2001
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.