missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium on activated carbon, 5% Ru, unreduced, ca. 50% moisture

Product Code. 10531261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10531261 100 g Glass bottle 100g
10521261 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10531261 Proveedor Thermo Scientific Acros N.º de proveedor 195461000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 7440-18-8 | Ru | 101.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-18-8 , 7440-44-0 , 7732-18-5
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Especificaciones

Color Black
Density 1.02
Quantity 100 g
Packaging Glass bottle
Form Powder
Fieser 01,983; 05,574
Merck Index 15, 8433
Formula Weight 101.07
Percent Purity 5% (Ru on dry basis)(typically)
Chemical Name or Material Ruthenium on activated carbon, 5% Ruthenium

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.