missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylchloride hydrate

Product Code. 11324748
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11324748

Brand: Thermo Scientific Alfa Aesar 043089.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32535-76-5 | Cl3NORu | 237.42 g/mol

Used as catalyst. As a precursor to other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32535-76-5
Molecular Formula Cl3NORu
Molecular Weight (g/mol) 237.42
MDL Number MFCD00049765
InChI Key XTHALOZYNGMAKY-UHFFFAOYSA-K
IUPAC Name trichloro(nitroso)rutheniumtris(ylium)
SMILES Cl[Ru+3](Cl)(Cl)N=O

Specifications

Melting Point 180°C (decomposition)
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 237.44 (Anhydrous)
Chemical Name or Material Ruthenium (III) nitrosylchloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.