missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ruthenium Red, ≥85%, pure

Product Code. 15519343 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15519343

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11103-72-3 | H24Cl6N14O2Ru3 | 786.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11103-72-3
Molecular Formula H24Cl6N14O2Ru3
Molecular Weight (g/mol) 786.35
InChI Key JQJSTVUROJELSR-UHFFFAOYSA-H
Synonym 6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride
PubChem CID 16218584
IUPAC Name azane;ruthenium(2+);hexachloride;dihydrate
SMILES N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2]

Specifications

Melting Point > 500.0°C
Quantity 1 g
Assay Percent Range ≥85%
Linear Formula [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
Merck Index 15,8434
Solubility Information Solubility in water: soluble.
Formula Weight 786.35
Percent Purity ≥85%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Ruthenium Red
Show More Show Less
compliance-icons
Product Identifier
  • Ruthenium Red
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.