missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium Red (Technical Grade), TRC

Product Code. 30417296
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417296 100 mg
30416224 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417296 Supplier Toronto Research Chemicals Supplier No. TRCR701860100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11103-72-3
Molecular Formula 6 Cl . O2 Ru3 . 14 H3 N
Molecular Weight (g/mol) 786.35
Synonym Ammoniated ruthenium oxychloride
SMILES N.N.N.N.N.N.N.N.N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2]O[Ru+2]O[Ru+2]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/6ClH.14H3N.2O.3Ru/h6*1H;14*1H3;;;;;/q;;;;;;;;;;;;;;;;;;;;;;3*+2/p-6
Formula Weight 785.8877
Chemical Name or Material Ruthenium red
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.