missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Ruxolitinib​

Product Code. 17190212
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17190212

Brand: STEMCELL Technologies 73402

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

JAK/STAT pathway inhibitor; Inhibits JAK1 and JAK2

  • Ruxolitinib is an ATP mimic that inhibits all Janus-associated kinase (JAK) family kinases with a preference for JAK-1 and JAK-2 over JAK-3 and TYK2 with IC50 values of 3.3, 2.8, 428 and 19 nM respectively (Verstovsek; Quintás-Cardama et al.)
  • Suppresses erythroid progenitor colony formation from peripheral blood mononuclear cells of polycythemia vera patients with the constitutively active JAK2 V617F mutation
  • Reduces malignant cell proliferation and decreases interleukin-6 and TNF-α signaling in a JAK2 V617F-driven mouse model of myeloproliferative disorder (Quintás-Cardama et al.)
  • Promotes hair regrowth in a mouse model of alopecia areata (Xing et al.)

Specifications

CAS 941678-49-5
Synonym INCB 018424, INC 424
Quantity 1 mg
Molecular Formula C17H18N6
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.