missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Ruxolitinib phosphate

Product Code. 18775846
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18775846 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18775846 Supplier R&D Systems™ Supplier No. 7048/50
  • ONLINE EXCLUSIVE PRICE
    347.86 €
    311.85 € / 50mg
    Save 36.01 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Phosphate salt of Ruxolitinib (Cat. No. 7064); potent and selective JAK1/JAK2 inhibitor

Ruxolitinib phosphate is a phosphate salt of Ruxolitinib (Cat. No. 7064). Potent and selective JAK1/JAK2 inhibitor; orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 R)-3-Cyclopentyl-3-[4-(7 H-pyrrolo[2,3-d]pyrimidin-4-yl)-1 H-pyrazol-1-yl]propanenitrile phosphate
CAS 1092939-17-7
Quantity 50 mg
Product Line Tocris
Target JAK Kinase Inhibitors
Molecular Formula C17H18N6.H3O4P
Formula Weight 2323
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.